COMH4211B236 Combi-Blocks, Inc.

tert-Butyl 4-(5-bromopyrimidine-2-carbonyl)piperazine-1-carboxylate

  • MDL number MFCD31613812
  • Cas Number : 1630807-75-8
  • Molecular Weight : 371.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4274

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4274
Availability details for
Quantity
  • Availability
  • Add to Cart