COMH4211B237 Combi-Blocks, Inc.

R-(2-Hydroxy-1-methyl-ethyl)-carbamic acid allyl ester

  • MDL number MFCD21735562
  • Cas Number : 1240490-25-8
  • Molecular Weight : 159.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4276

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4276
Availability details for
Quantity
  • Availability
  • Add to Cart