COMH4211B238 Combi-Blocks, Inc.

4'-Chloro-4-methoxy-2'-methylbiphenyl-3-carbaldehyde

  • MDL number MFCD31583174
  • Cas Number : 408372-61-2
  • Molecular Weight : 260.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4277

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4277
Availability details for
Quantity
  • Availability
  • Add to Cart