COMH4211B239 Combi-Blocks, Inc.

2-((5-Bromo-2-chloropyrimidin-4-yl)amino)-n-methylbenzenesulfonamide

  • MDL number MFCD26399070
  • Cas Number : 761440-01-1
  • Molecular Weight : 377.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4279

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4279
Availability details for
Quantity
  • Availability
  • Add to Cart