COMH4211B23A Combi-Blocks, Inc.

(2-Isopropylsulfanyl-5-nitro-benzyl)-methyl-amine

  • MDL number MFCD31561539
  • Cas Number : 1037082-36-2
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4281

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4281
Availability details for
Quantity
  • Availability
  • Add to Cart