COMH4211B23B Combi-Blocks, Inc.

1-(3,4-Difluorobenzyl)-1h-imidazole-4-carboxylic acid

  • MDL number MFCD29479462
  • Cas Number : 1972201-13-0
  • Molecular Weight : 238.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4285

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4285
Availability details for
Quantity
  • Availability
  • Add to Cart