COMH4211B23C Combi-Blocks, Inc.

tert-Butyl 4-((5-bromo-1h-indol-1-yl)methyl)piperidine-1-carboxylate

  • MDL number MFCD31561614
  • Cas Number : 1808957-13-2
  • Molecular Weight : 393.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4287

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4287
Availability details for
Quantity
  • Availability
  • Add to Cart