COMH4211B23E Combi-Blocks, Inc.

1-(3-Chloro-2-ethoxyphenyl)ethan-1-amine

  • MDL number MFCD16304818
  • Cas Number : 1225522-64-4
  • Molecular Weight : 199.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4295

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4295
Availability details for
Quantity
  • Availability
  • Add to Cart