COMH4211B23F Combi-Blocks, Inc.

4-Amino-2-(2-methoxy-ethoxy)-n,n-dimethyl-benzamide

  • MDL number MFCD19078933
  • Cas Number : 1466071-92-0
  • Molecular Weight : 238.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4298

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4298
Availability details for
Quantity
  • Availability
  • Add to Cart