COMH4211B240 Combi-Blocks, Inc.

1-(3-Fluoro-2-methyl-phenyl)-propylamine hydrochloride

  • MDL number MFCD31557785
  • Cas Number : 2204052-12-8
  • Molecular Weight : 203.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-fluoro-2-methylphenyl)propan-1-amine hydrochloride
Inchi Code 1S/C10H14FN.ClH/c1-3-10(12)8-5-4-6-9(11)7(8)2;/h4-6,10H,3,12H2,1-2H3;1H
Inchi Key YWWOBCHCJIPJKF-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4304

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4304
Availability details for
Quantity
  • Availability
  • Add to Cart