COMH4211B241 Combi-Blocks, Inc.

5-Bromo-2-methoxy-n,n-dimethylnicotinamide

  • MDL number MFCD26129078
  • Cas Number : 1072854-96-6
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4305

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4305
Availability details for
Quantity
  • Availability
  • Add to Cart