COMH4211B242 Combi-Blocks, Inc.

2-[2-(2-2-[2-(tert-Butyl-dimethyl-silanyloxy)-ethoxy]-ethoxy-ethoxy)-ethoxy]-ethanol

  • MDL number MFCD31614078
  • Cas Number : 393174-68-0
  • Molecular Weight : 352.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4310

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4310
Availability details for
Quantity
  • Availability
  • Add to Cart