COMH4211B243 Combi-Blocks, Inc.

4-(4-Methoxy-pyrazol-1-ylmethyl)-benzoic acid

  • MDL number MFCD31561529
  • Cas Number : 2018287-95-9
  • Molecular Weight : 232.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4311

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4311
Availability details for
Quantity
  • Availability
  • Add to Cart