COMH4211B244 Combi-Blocks, Inc.

(4-(Pyridin-2-ylmethoxy)-phenyl)-methanol

  • MDL number MFCD11149436
  • Cas Number : 50596-34-4
  • Molecular Weight : 215.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4312

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4312
Availability details for
Quantity
  • Availability
  • Add to Cart