COMH4211B247 Combi-Blocks, Inc.

1-(4-Chloro-benzyl)-piperidin-4-ol

  • MDL number MFCD00976663
  • Cas Number : 21937-62-2
  • Molecular Weight : 225.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4318

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4318
Availability details for
Quantity
  • Availability
  • Add to Cart