COMH4211B24D Combi-Blocks, Inc.

1-(4-Isobutoxy-2-methyl-phenyl)-ethylamine hydrochloride

  • MDL number MFCD31557832
  • Cas Number : 2203139-81-3
  • Molecular Weight : 243.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4334

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4334
Availability details for
Quantity
  • Availability
  • Add to Cart