COMH4211B24E Combi-Blocks, Inc.

N-Benzyl-5-hydroxycyclohex-3-enecarboxamide

  • MDL number MFCD23703085
  • Cas Number : 61088-56-0
  • Molecular Weight : 231.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4335

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4335
Availability details for
Quantity
  • Availability
  • Add to Cart