COMH4211B24F Combi-Blocks, Inc.

2-Methyl-4-piperidin-4-yl-benzoic acid methyl ester

  • MDL number MFCD31583153
  • Cas Number : 2228406-31-1
  • Molecular Weight : 233.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-methyl-4-(piperidin-4-yl)benzoate
Inchi Code 1S/C14H19NO2/c1-10-9-12(11-5-7-15-8-6-11)3-4-13(10)14(16)17-2/h3-4,9,11,15H,5-8H2,1-2H3
Inchi Key UQKRMGBLCNKWGA-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4336

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4336
Availability details for
Quantity
  • Availability
  • Add to Cart