COMH4211B250 Combi-Blocks, Inc.

Cyclopropyl-(4-methoxy-2-trifluoromethoxy-benzyl)-amine

  • MDL number MFCD31614075
  • Cas Number : 2270906-97-1
  • Molecular Weight : 261.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(4-methoxy-2-(trifluoromethoxy)benzyl)cyclopropanamine
Inchi Code 1S/C12H14F3NO2/c1-17-10-5-2-8(7-16-9-3-4-9)11(6-10)18-12(13,14)15/h2,5-6,9,16H,3-4,7H2,1H3
Inchi Key KKOASGLPRDDHIR-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4337

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4337
Availability details for
Quantity
  • Availability
  • Add to Cart