COMH4211B253 Combi-Blocks, Inc.

3-Bromo-5-chloro-n,n-dimethylbenzamide

  • MDL number MFCD26611405
  • Cas Number : 1369869-37-3
  • Molecular Weight : 262.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4343

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4343
Availability details for
Quantity
  • Availability
  • Add to Cart