COMH4211B256 Combi-Blocks, Inc.

5-(tert-Butoxycarbonylamino)-1-methyl-1h-pyrazole-4-carboxylic acid

  • MDL number MFCD16875652
  • Cas Number : 916850-80-1
  • Molecular Weight : 241.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4349

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4349
Availability details for
Quantity
  • Availability
  • Add to Cart