COMH4211B258 Combi-Blocks, Inc.

4-Bromo-1-methyl-2-(2-methylpropoxy)benzene

  • MDL number MFCD18917816
  • Cas Number : 1309933-19-4
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4351

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4351
Availability details for
Quantity
  • Availability
  • Add to Cart