COMH4211B259 Combi-Blocks, Inc.

1-Iodo-4-(2-methoxyethoxy)benzene

  • MDL number MFCD14647006
  • Cas Number : 102293-99-2
  • Molecular Weight : 278.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4353

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4353
Availability details for
Quantity
  • Availability
  • Add to Cart