COMH4211B25A Combi-Blocks, Inc.

Carbamic acid, n-[(1s)-1-(4-chloro-2-pyridinyl)-3-buten-1-yl]-, 1,1-dimethylethyl ester

  • MDL number MFCD29076589
  • Cas Number : 1329171-69-8
  • Molecular Weight : 282.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4356

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4356
Availability details for
Quantity
  • Availability
  • Add to Cart