COMH4211B25D Combi-Blocks, Inc.

2-Chloro-n-(4-cyclopropylmethyl amino)-n-(6-n-propylamino) [1,3,5]triazine

  • MDL number MFCD23983113
  • Cas Number : 40533-54-8
  • Molecular Weight : 241.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4360

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4360
Availability details for
Quantity
  • Availability
  • Add to Cart