COMH4211B25E Combi-Blocks, Inc.

1-Allyl-1h-indole-2-carboxylic acid

  • MDL number MFCD11155425
  • Cas Number : 155193-48-9
  • Molecular Weight : 201.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4362

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4362
Availability details for
Quantity
  • Availability
  • Add to Cart