COMH4211B25F Combi-Blocks, Inc.

Benzoic acid, 2-hydroxy-5-(2-propyn-1-yloxy)-, methyl ester

  • MDL number MFCD24724325
  • Cas Number : 73922-94-8
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4363

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4363
Availability details for
Quantity
  • Availability
  • Add to Cart