COMH4211B260 Combi-Blocks, Inc.

Toluene-4-sulfonic acid 2-(2-(2-[2-(tert-butyl-dimethyl-silanyloxy)-ethoxy]-ethoxy)-ethoxy)-ethyl ester

  • MDL number MFCD31381597
  • Cas Number : 571167-64-1
  • Molecular Weight : 462.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4365

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4365
Availability details for
Quantity
  • Availability
  • Add to Cart