COMH4211B262 Combi-Blocks, Inc.

3-Fluoro-5-(propan-2-yloxy)benzoic acid

  • MDL number MFCD28356762
  • Cas Number : 1369879-36-6
  • Molecular Weight : 198.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4368

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4368
Availability details for
Quantity
  • Availability
  • Add to Cart