COMH4211B264 Combi-Blocks, Inc.

(2-Bromo-3-ethoxyphenyl)methanamine

  • MDL number MFCD31613857
  • Cas Number : 1934458-37-3
  • Molecular Weight : 230.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-bromo-3-ethoxyphenyl)methanamine
Inchi Code 1S/C9H12BrNO/c1-2-12-8-5-3-4-7(6-11)9(8)10/h3-5H,2,6,11H2,1H3
Inchi Key NIZAKGQEGXTVHJ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4370

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4370
Availability details for
Quantity
  • Availability
  • Add to Cart