COMH4211B265 Combi-Blocks, Inc.

2,6-Dichloro-9-ethyl-9h-purine

  • MDL number MFCD11047296
  • Cas Number : 190655-14-2
  • Molecular Weight : 217.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4373

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4373
Availability details for
Quantity
  • Availability
  • Add to Cart