COMH4211B285 Combi-Blocks, Inc.

4-Hydroxy-2-methyl-n-propylbenzamide

  • MDL number MFCD31613838
  • Cas Number : 1695767-59-9
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4382

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4382
Availability details for
Quantity
  • Availability
  • Add to Cart