COMH4211B286 Combi-Blocks, Inc.

2-[2-Fluoro-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenyl]-n,n-dimethyl-acetamide

  • MDL number MFCD18760162
  • Molecular Weight : 307.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4386

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4386
Availability details for
Quantity
  • Availability
  • Add to Cart