COMH4211B287 Combi-Blocks, Inc.

4-Amino-2-(2-methoxyethoxy)-n-methylbenzamide

  • MDL number MFCD19640848
  • Cas Number : 1340427-29-3
  • Molecular Weight : 224.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4390

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4390
Availability details for
Quantity
  • Availability
  • Add to Cart