COMH4211B288 Combi-Blocks, Inc.

Desloratadine pyridine n-oxide

  • MDL number MFCD11977840
  • Cas Number : 169253-26-3
  • Molecular Weight : 326.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4391

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4391
Availability details for
Quantity
  • Availability
  • Add to Cart