COMH4211B28A Combi-Blocks, Inc.

1-(3-Chloropropyl)-3-(4-nitrophenyl)urea

  • MDL number MFCD27933639
  • Cas Number : 936112-75-3
  • Molecular Weight : 257.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4397

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4397
Availability details for
Quantity
  • Availability
  • Add to Cart