COMH4211B28B Combi-Blocks, Inc.

4-(4-Ethoxyphenyl)butan-2-one

  • MDL number MFCD09942355
  • Cas Number : 86416-81-1
  • Molecular Weight : 192.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4398

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4398
Availability details for
Quantity
  • Availability
  • Add to Cart