COMH4211B28D Combi-Blocks, Inc.

2,5-Dioxopyrrolidin-1-yl 3-((2-fluorobenzyl)oxy)propanoate

  • MDL number MFCD20226177
  • Cas Number : 1134280-64-0
  • Molecular Weight : 295.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4399

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4399
Availability details for
Quantity
  • Availability
  • Add to Cart