COMH4211B290 Combi-Blocks, Inc.

N-Alloc-1,4-butandiamine hydrochloride

  • MDL number MFCD06798160
  • Cas Number : 1049722-10-2
  • Molecular Weight : 208.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4402

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4402
Availability details for
Quantity
  • Availability
  • Add to Cart