COMH4211B292 Combi-Blocks, Inc.

4-Cyclopropylmethoxy-2-methoxy-benzylamine hydrochloride

  • MDL number MFCD31539578
  • Cas Number : 2203194-94-7
  • Molecular Weight : 243.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4409

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4409
Availability details for
Quantity
  • Availability
  • Add to Cart