COMH4211B293 Combi-Blocks, Inc.

N-Ethyl-4-hydroxy-2-methylbenzamide

  • MDL number MFCD30087657
  • Cas Number : 1695433-02-3
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4410

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4410
Availability details for
Quantity
  • Availability
  • Add to Cart