COMH4211B294 Combi-Blocks, Inc.

2',4'-Difluoro-[1,1'-biphenyl]-3-amine

  • MDL number MFCD04117391
  • Cas Number : 160521-67-5
  • Molecular Weight : 205.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4413

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4413
Availability details for
Quantity
  • Availability
  • Add to Cart