COMH4211B295 Combi-Blocks, Inc.

1-[5-Methyl-2-(propan-2-yloxy)phenyl]methanamine

  • MDL number MFCD18331107
  • Cas Number : 1341803-00-6
  • Molecular Weight : 179.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4415

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4415
Availability details for
Quantity
  • Availability
  • Add to Cart