COMH4211B298 Combi-Blocks, Inc.

Cyclopropyl-(6-methoxy-pyridin-3-ylmethyl)-amine

  • MDL number MFCD16704260
  • Cas Number : 1249037-91-9
  • Molecular Weight : 178.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4425

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4425
Availability details for
Quantity
  • Availability
  • Add to Cart