COMH4211B299 Combi-Blocks, Inc.

6-Bromo-1-(2-difluoromethoxybenzyl)-2-methyl-1h-benzimidazole

  • MDL number MFCD31579551
  • Cas Number : 1515921-82-0
  • Molecular Weight : 367.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4426

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4426
Availability details for
Quantity
  • Availability
  • Add to Cart