COMH4211B29A Combi-Blocks, Inc.

3-Fluoro-4-(1-isobutyl-piperidin-4-yl)-phenol

  • MDL number MFCD31583401
  • Cas Number : 2270909-48-1
  • Molecular Weight : 251.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4428

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4428
Availability details for
Quantity
  • Availability
  • Add to Cart