COMH4211B29C Combi-Blocks, Inc.

[3-(5-Chloro-pyridin-2-yl)-5-methyl-isoxazol-4-yl]-methanol

  • MDL number MFCD31579559
  • Cas Number : 1159252-17-1
  • Molecular Weight : 224.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4436

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4436
Availability details for
Quantity
  • Availability
  • Add to Cart