COMH4211B29E Combi-Blocks, Inc.

4-Piperidin-4-yl-2-trifluoromethyl-benzoic acid methyl ester

  • MDL number MFCD31583155
  • Molecular Weight : 287.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4438

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4438
Availability details for
Quantity
  • Availability
  • Add to Cart