COMH4211B2A0 Combi-Blocks, Inc.

3-(4-Nitro-phenoxycarbonyloxymethyl)-benzoic acid methyl ester

  • MDL number MFCD31583358
  • Cas Number : 2270909-29-8
  • Molecular Weight : 331.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4442

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4442
Availability details for
Quantity
  • Availability
  • Add to Cart