COMH4211B2A1 Combi-Blocks, Inc.

3-Cyclopentylmethyl-benzoic acid

  • MDL number MFCD31614029
  • Cas Number : 1783663-54-6
  • Molecular Weight : 204.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4447

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4447
Availability details for
Quantity
  • Availability
  • Add to Cart